Gene Transcripts

Gene Symbol
REXO4
Entrez Gene ID
57109
Gene Name
REX4 homolog, 3'-5' exonuclease
Gene Aliases
REX4, XPMC2, XPMC2H
Chromosome Location
Chr. 9: 133406058 - 133418172 on Build GRCh38
UniGene ID
Hs.438409
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001279350.1 NP_001266279.1 3 723
NM_001279351.1 NP_001266280.1 3 584
NM_020385.3 NP_065118.2 3 901
NR_103995.1 2 554
NR_103996.1 2 554
GenBank mRNA AF273304.1 - 3 849
AF295774.1 - 3 849
AK025493.1 - 3 755
AK295982.1 - 3 584
AK304547.1 - 3 639
AK310164.1 - 2 457
AK314333.1 - 3 815
AK315904.1 - 4 574
AL136894.1 - 3 832
BC009274.2 - 3 804 3958331
BC071691.1 - 2 413

View Full Product Details