Gene Transcripts

Gene Symbol
AAK1
Entrez Gene ID
22848
Gene Name
AP2 associated kinase 1
Gene Aliases
Chromosome Location
Chr. 2: 69457995 - 69643845 on Build GRCh38
UniGene ID
Hs.468878
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014911.3 NP_055726.3 15 2548
GenBank mRNA AB028971.1 - 15 2427
BC104842.1 - 14 2067 8143845
BC111965.1 - 14 2067 8327433
BC143710.1 - 14 2067 9052224

View Full Product Details