Gene Transcripts
- Gene Symbol
- PTTG1
- Entrez Gene ID
- 9232
- Gene Name
- pituitary tumor-transforming 1
- Gene Aliases
- EAP1, HPTTG, PTTG, TUTR1
- Chromosome Location
- Chr. 5: 160421807 - 160428744 on Build GRCh38
- UniGene ID
- Hs.350966
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001282382.1 |
NP_001269311.1
|
3, 4 |
768 |
|
NM_001282383.1 |
NP_001269312.1
|
4, 5 |
622 |
|
NM_004219.3 |
NP_004210.1
|
4, 5 |
461 |
|
GenBank mRNA |
AF062649.1 |
- |
4, 5 |
432 |
|
AF075242.1 |
- |
3, 4 |
385 |
|
AF095287.1 |
- |
4, 5 |
401 |
|
AJ223953.1 |
- |
3, 4 |
470 |
|
AK312209.1 |
- |
4, 5 |
439 |
|
BC101834.1 |
- |
4, 5 |
394 |
8069340 |
BC108894.1 |
- |
3, 4 |
365 |
|
BC108895.1 |
- |
3, 4 |
365 |
|
BC112071.1 |
- |
4, 5 |
394 |
8327539 |
BC113842.1 |
- |
3, 4 |
365 |
|
BC113896.1 |
- |
3, 4 |
365 |
|
BC128193.1 |
- |
3, 4 |
365 |
40113885 |
BC128194.1 |
- |
3, 4 |
365 |
40113886 |
BU182149.1 |
- |
4, 5 |
617 |
|
CR457135.1 |
- |
3, 4 |
365 |
|
CR541685.1 |
- |
3, 4 |
365 |
|
HY006026.1 |
- |
4, 5 |
463 |
|
View Full Product Details