Gene Transcripts

Gene Symbol
FNDC3A
Entrez Gene ID
22862
Gene Name
fibronectin type III domain containing 3A
Gene Aliases
FNDC3, HUGO, bA203I16.1, bA203I16.5
Chromosome Location
Chr. 13: 48975241 - 49209779 on Build GRCh38
UniGene ID
Hs.508010
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001079673.1 NP_001073141.1 23, 24 3286
NM_001278438.1 NP_001265367.1 23, 24 3329
NM_014923.4 NP_055738.3 21, 22 2910
NR_103528.1 23, 24 3318
XM_011534997.2 XP_011533299.1 22, 23 3000
XM_017020440.1 XP_016875929.1 23, 24 3171
XM_017020441.1 XP_016875930.1 25, 26 3792
XM_017020442.1 XP_016875931.1 24, 25 3324
GenBank mRNA AB023187.2 - 21, 22 2840
AJ749706.1 - 23, 24 3288
AJ749707.1 - 21, 22 2901
AK172814.1 - 1, 2 217
AK302415.1 - 21, 22 2910
BC060816.1 - 14, 15 1948
BC070072.1 - 14, 15 1948
BC132812.1 - 23, 24 3053 40146834
BC136617.1 - 23, 24 3054 9020607
BC144301.1 - 23, 24 3054 9052821
BX640739.1 - 23, 24 3189
BX648141.1 - 23, 24 3276
BX648706.1 - 25, 26 3134

View Full Product Details