Gene Transcripts

Gene Symbol
MON1B
Entrez Gene ID
22879
Gene Name
MON1 homolog B, secretory trafficking associated
Gene Aliases
HSRG1, SAND2, SRG1
Chromosome Location
Chr. 16: 77190919 - 77199646 on Build GRCh38
UniGene ID
Hs.513743
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286639.1 NP_001273568.1 5 1533
NM_001286640.1 NP_001273569.1 4 1375
NM_014940.3 NP_055755.1 6 1860
GenBank mRNA AB020679.1 - 6 1544
AF442486.1 - 5 1524
AK023374.1 - 5 801
AK023896.1 - 6 1588
AK128411.1 - 3 1402
AK293392.1 - 4 1095
AK296467.1 - 5 1260
BC024277.1 - 6 1840 4843859

View Full Product Details