Gene Transcripts

Gene Symbol
SMG5
Entrez Gene ID
23381
Gene Name
SMG5, nonsense mediated mRNA decay factor
Gene Aliases
EST1B, LPTS-RP1, LPTSRP1, SMG-5
Chromosome Location
Chr. 1: 156249224 - 156291498 on Build GRCh38
UniGene ID
Hs.516837
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001323614.1 NP_001310543.1 12 1749
NM_001323615.1 NP_001310544.1 11 1611
NM_001323616.1 NP_001310545.1 13 1881
NM_001323617.1 NP_001310546.1 13 1716
NM_015327.2 NP_056142.2 12 1688
XM_017000842.1 XP_016856331.1 8 1634
XM_017000843.1 XP_016856332.1 11 1616
GenBank mRNA AB029012.2 - 12 1690
AB085691.1 - 12 1616
AY168922.1 - 12 1542
AY336728.1 - 12 1539
BC007453.2 - 1 135
BC038296.1 - 12 1689 4824487
CR933677.1 - 1 67

View Full Product Details