Gene Transcripts
- Gene Symbol
- LCE5A
- Entrez Gene ID
- 254910
- Gene Name
- late cornified envelope 5A
- Gene Aliases
- LEP18, SPRL5A
- Chromosome Location
- Chr. 1: 152510844 - 152512177 on Build GRCh38
- UniGene ID
- Hs.516410
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_178438.4 |
NP_848525.1
|
2 |
632 |
|
View Full Product Details