Gene Transcripts

Gene Symbol
BAHD1
Entrez Gene ID
22893
Gene Name
bromo adjacent homology domain containing 1
Gene Aliases
Chromosome Location
Chr. 15: 40439721 - 40468242 on Build GRCh38
UniGene ID
Hs.22109
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301132.1 NP_001288061.1 2 891
NM_014952.4 NP_055767.3 2 764
XM_011521366.2 XP_011519668.2 2 888
XM_011521367.2 XP_011519669.2 2 893
XM_011521368.2 XP_011519670.2 2 887
XM_011521369.2 XP_011519671.1 2 742
GenBank mRNA AB023162.1 - 2 891
BC022782.2 - 2 703 3903581

View Full Product Details