Gene Transcripts

Gene Symbol
RPH3A
Entrez Gene ID
22895
Gene Name
rabphilin 3A
Gene Aliases
Chromosome Location
Chr. 12: 112791538 - 112898881 on Build GRCh38
UniGene ID
Hs.21239
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001143854.1 NP_001137326.1 20, 21 2274
NM_014954.3 NP_055769.2 19, 20 2262
XM_011538048.1 XP_011536350.1 20, 21 2078
XM_017019034.1 XP_016874523.1 12, 13 1532
GenBank mRNA AB023202.1 - 20, 21 2195
AK295696.1 - 20, 21 2228
AK296296.1 - 18, 19 1813
AK316112.1 - 20, 21 2109
BC017259.2 - 19, 20 2069 3510158
BX647281.1 - 12, 13 2868

View Full Product Details