Gene Transcripts

Gene Symbol
KRTAP5-10
Entrez Gene ID
387273
Gene Name
keratin associated protein 5-10
Gene Aliases
KRTAP5.10
Chromosome Location
Chr. 11: 71565563 - 71566620 on Build GRCh38
UniGene ID
Hs.661887
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012710.1 NP_001012728.1 1 1016
GenBank mRNA AB126079.1 - 1 1016

View Full Product Details