Gene Transcripts

Gene Symbol
TARM1
Entrez Gene ID
441864
Gene Name
T cell-interacting, activating receptor on myeloid cells 1
Gene Aliases
OLT-2
Chromosome Location
Chr. 19: 54069342 - 54081365 on Build GRCh38
UniGene ID
Hs.694674
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135686.2 NP_001129158.2 5 701
XM_005278278.3 XP_005278335.1 5 970
GenBank mRNA AK301730.1 - 5 986
DQ479398.1 - 5 701

View Full Product Details