Gene Transcripts
- Gene Symbol
- CCT5
- Entrez Gene ID
- 22948
- Gene Name
- chaperonin containing TCP1 subunit 5
- Gene Aliases
- CCT-epsilon, CCTE, HEL-S-69, PNAS-102, TCP-1-epsilon
- Chromosome Location
- Chr. 5: 10249921 - 10266412 on Build GRCh38
- UniGene ID
- Hs.1600
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001306153.1 |
NP_001293082.1
|
2, 3 |
189 |
|
NM_001306156.1 |
NP_001293085.1
|
2, 3 |
421 |
|
NM_012073.4 |
NP_036205.1
|
2, 3 |
589 |
|
GenBank mRNA |
AK289353.1 |
- |
2, 3 |
187 |
|
AK293341.1 |
- |
2, 3 |
258 |
|
AK301760.1 |
- |
2, 3 |
181 |
|
AK301985.1 |
- |
2, 3 |
228 |
|
AK303086.1 |
- |
2, 3 |
218 |
|
AK316376.1 |
- |
2, 3 |
176 |
|
BC002971.1 |
- |
2, 3 |
164 |
|
BC006543.2 |
- |
2, 3 |
204 |
2824199 |
BC035499.2 |
- |
2, 3 |
251 |
4329194 |
BC040027.1 |
- |
2, 3 |
221 |
|
BU858349.1 |
- |
2, 3 |
357 |
|
D43950.2 |
- |
2, 3 |
206 |
|
DB449481.1 |
- |
|
422 |
|
DC395133.1 |
- |
2, 3 |
181 |
|
FJ224303.1 |
- |
2, 3 |
340 |
|
View Full Product Details