Gene Transcripts

Gene Symbol
PDZK1
Entrez Gene ID
5174
Gene Name
PDZ domain containing 1
Gene Aliases
CAP70, CLAMP, NHERF-3, NHERF3, PDZD1
Chromosome Location
Chr. 1: 145670852 - 145707507 on Build GRCh38
UniGene ID
Hs.444751
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001201325.1 NP_001188254.1 8 1484
NM_001201326.1 NP_001188255.1 6 1151
NM_002614.4 NP_002605.2 9 1555
XM_011509616.2 XP_011507918.1 7 1400
XM_011509618.2 XP_011507920.1 8 1488
XM_011509619.2 XP_011507921.2 9 1513
XM_017001464.1 XP_016856953.1 7 1400
XM_017001465.1 XP_016856954.1 9 1513
XM_017001466.1 XP_016856955.1 9 1513
XM_017001467.1 XP_016856956.1 7 1454
GenBank mRNA AF012281.1 - 8 1431
AK298269.1 - 6 1092
AK313100.1 - 8 1436
BC006496.2 - 8 1495 2959384
BC006518.2 - 8 1495 2959384

Gene Symbol
PDZK1P1
Entrez Gene ID
100034743
Gene Name
PDZ domain containing 1 pseudogene 1
Gene Aliases
OTTHUMT00000038524, PDZK1P2
Chromosome Location
Chr. 1: 147993862 - 148014956 on Build GRCh38
UniGene ID
Hs.696575
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_111936.1 9 1528
GenBank mRNA AK126719.1 - 8 2897

View Full Product Details