Gene Transcripts
- Gene Symbol
- UCA1
- Entrez Gene ID
- 652995
- Gene Name
- urothelial cancer associated 1 (non-protein coding)
- Gene Aliases
- CUDR, LINC00178, NCRNA00178, UCAT1, onco-lncRNA-36
- Chromosome Location
- Chr. 19: 15828947 - 15836321 on Build GRCh38
- UniGene ID
- Hs.644234
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_015379.3 |
|
3 |
1770 |
|
GenBank mRNA |
AK092401.1 |
- |
3 |
1756 |
|
GU357550.1 |
- |
3 |
1622 |
|
View Full Product Details