Gene Transcripts
- Gene Symbol
- CGNL1
- Entrez Gene ID
- 84952
- Gene Name
- cingulin like 1
- Gene Aliases
- JACOP, PCING
- Chromosome Location
- Chr. 15: 57376487 - 57550727 on Build GRCh38
- UniGene ID
- Hs.148989
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001252335.1 |
NP_001239264.1
|
6, 7 |
2106 |
|
NM_032866.4 |
NP_116255.2
|
5, 6 |
1991 |
|
XM_005254726.3 |
XP_005254783.1
|
5, 6 |
2009 |
|
XM_005254727.4 |
XP_005254784.1
|
6, 7 |
2387 |
|
XM_011522120.1 |
XP_011520422.1
|
6, 7 |
2045 |
|
XM_011522121.1 |
XP_011520423.1
|
5, 6 |
2168 |
|
XM_017022685.1 |
XP_016878174.1
|
5, 6 |
2626 |
|
XM_017022686.1 |
XP_016878175.1
|
5, 6 |
2624 |
|
GenBank mRNA |
AB051536.2 |
- |
4, 5 |
1150 |
|
AK056673.1 |
- |
5, 6 |
1991 |
|
AK307410.1 |
- |
5, 6 |
1991 |
|
AY274808.1 |
- |
5, 6 |
1989 |
|
AY610514.1 |
- |
6, 7 |
2086 |
|
BC093827.1 |
- |
4, 5 |
1933 |
7939672 |
BC112049.1 |
- |
4, 5 |
1945 |
8327517 |
BC118918.1 |
- |
4, 5 |
1932 |
40074275 |
View Full Product Details