Gene Transcripts
- Gene Symbol
- NUBP2
- Entrez Gene ID
- 10101
- Gene Name
- nucleotide binding protein 2
- Gene Aliases
- CFD1, NBP 2, NUBP1
- Chromosome Location
- Chr. 16: 1782923 - 1789191 on Build GRCh38
- UniGene ID
- Hs.256549
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001284501.1 |
NP_001271430.1
|
4, 5 |
625 |
|
NM_012225.3 |
NP_036357.1
|
3, 4 |
435 |
|
XM_005255027.2 |
XP_005255084.1
|
3, 4 |
902 |
|
XM_017022831.1 |
XP_016878320.1
|
3, 4 |
569 |
|
GenBank mRNA |
AF118394.1 |
- |
3, 4 |
400 |
|
AK001023.1 |
- |
3, 4 |
411 |
|
AK315611.1 |
- |
3, 4 |
376 |
|
BC002768.2 |
- |
3, 4 |
392 |
3633393 |
BC008005.1 |
- |
3, 4 |
417 |
3507768 |
BU189242.1 |
- |
3, 4 |
435 |
|
BX446436.2 |
- |
4, 5 |
561 |
|
View Full Product Details