Gene Transcripts
- Gene Symbol
- NBPF8
- Entrez Gene ID
- 728841
- Gene Name
- neuroblastoma breakpoint family member 8
- Gene Aliases
- NBPF8P
- Chromosome Location
- Chr. 1: 120436351 - 120467844 on Build GRCh38
- UniGene ID
- Hs.445080
- Species
- Homo sapiens
- Gene Symbol
- NBPF1
- Entrez Gene ID
- 55672
- Gene Name
- neuroblastoma breakpoint family member 1
- Gene Aliases
- AB13, AB14, AB23, AD2, NBG, NBPF
- Chromosome Location
- Chr. 1: 16562427 - 16613605 on Build GRCh38
- UniGene ID
- Hs.467587
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC094705.1 |
- |
1 |
33 |
|
- Gene Symbol
- NBPF15
- Entrez Gene ID
- 284565
- Gene Name
- neuroblastoma breakpoint family member 15
- Gene Aliases
- AB14, AG3, NBPF16
- Chromosome Location
- Chr. 1: 144421386 - 144459464 on Build GRCh38
- UniGene ID
- Hs.523572
- Species
- Homo sapiens
- Gene Symbol
- NBPF26
- Entrez Gene ID
- 101060684
- Gene Name
- neuroblastoma breakpoint family member 26
- Gene Aliases
- Chromosome Location
- Chr. 1: 120805561 - 120842215 on Build GRCh38
- UniGene ID
- Hs.534675
- Species
- Homo sapiens
View Full Product Details