Gene Transcripts
- Gene Symbol
- GRAMD4
- Entrez Gene ID
- 23151
- Gene Name
- GRAM domain containing 4
- Gene Aliases
- DIP, dA59H18.1, dJ439F8.1
- Chromosome Location
- Chr. 22: 46576017 - 46682755 on Build GRCh38
- UniGene ID
- Hs.475150
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_015124.4 |
NP_055939.1
|
11, 12 |
1147 |
|
XM_005261396.3 |
XP_005261453.1
|
11, 12 |
1083 |
|
XM_006724169.1 |
XP_006724232.1
|
11, 12 |
1045 |
|
XM_006724170.3 |
XP_006724233.1
|
11, 12 |
1064 |
|
XM_011530011.1 |
XP_011528313.1
|
10, 11 |
983 |
|
XM_011530012.1 |
XP_011528314.1
|
10, 11 |
983 |
|
XM_011530013.2 |
XP_011528315.2
|
11, 12 |
1092 |
|
XM_011530014.2 |
XP_011528316.1
|
10, 11 |
983 |
|
XM_017028670.1 |
XP_016884159.1
|
11, 12 |
1105 |
|
GenBank mRNA |
AB018310.2 |
- |
11, 12 |
1147 |
|
BC129837.1 |
- |
10, 11 |
968 |
40119070 |
CR456470.1 |
- |
10, 11 |
973 |
|
View Full Product Details