Gene Transcripts

Gene Symbol
Cyp2d11
Entrez Gene ID
545123
Gene Name
cytochrome P450, family 2, subfamily d, polypeptide 11
Gene Aliases
Cyp2d, P450-2D
Chromosome Location
Chr. 15: 82389154 - 82394022 on Build GRCm38
UniGene ID
Mm.482764
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001104531.1 NP_001098001.1 7 1234

View Full Product Details