Gene Transcripts
- Gene Symbol
- RFTN1
- Entrez Gene ID
- 23180
- Gene Name
- raftlin, lipid raft linker 1
- Gene Aliases
- MIG2, PIB10, PIG9, RAFTLIN
- Chromosome Location
- Chr. 3: 16315845 - 16513715 on Build GRCh38
- UniGene ID
- Hs.98910
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_015150.1 |
NP_055965.1
|
5, 6 |
1113 |
|
XM_005264985.1 |
XP_005265042.1
|
5, 6 |
1070 |
|
XM_005264986.2 |
XP_005265043.1
|
5, 6 |
967 |
|
XM_006713069.1 |
XP_006713132.1
|
5, 6 |
976 |
|
XM_011533529.1 |
XP_011531831.1
|
7, 8 |
1510 |
|
XM_011533530.1 |
XP_011531832.1
|
5, 6 |
1138 |
|
XM_011533531.1 |
XP_011531833.1
|
7, 8 |
1510 |
|
XM_017006005.1 |
XP_016861494.1
|
7, 8 |
1510 |
|
XM_017006006.1 |
XP_016861495.1
|
7, 8 |
1510 |
|
XM_017006007.1 |
XP_016861496.1
|
5, 6 |
1097 |
|
GenBank mRNA |
AK074446.1 |
- |
5, 6 |
919 |
|
AK301024.1 |
- |
4, 5 |
806 |
|
AY237654.1 |
- |
5, 6 |
1104 |
|
BC006400.2 |
- |
1, 2 |
41 |
|
BC014032.2 |
- |
1, 2 |
41 |
|
BC032349.1 |
- |
4, 5 |
786 |
|
BC051336.1 |
- |
5, 6 |
890 |
|
BC069209.1 |
- |
5, 6 |
890 |
|
BC098270.1 |
- |
5, 6 |
937 |
40001191 |
BC100667.1 |
- |
5, 6 |
937 |
40001188 |
BC100669.1 |
- |
5, 6 |
937 |
40001190 |
BC121795.1 |
- |
5, 6 |
937 |
40011545 |
D42043.1 |
- |
5, 6 |
1041 |
|
View Full Product Details