Gene Transcripts
- Gene Symbol
- CYFIP1
- Entrez Gene ID
- 23191
- Gene Name
- cytoplasmic FMR1 interacting protein 1
- Gene Aliases
- P140SRA-1, SHYC, SRA-1, SRA1
- Chromosome Location
- Chr. 15: 22869465 - 22980906 on Build GRCh38
- UniGene ID
- Hs.26704
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001033028.1 |
NP_001028200.1
|
5, 6 |
1285 |
|
NM_001287810.2 |
NP_001274739.1
|
21, 22 |
2556 |
|
NM_001324119.1 |
NP_001311048.1
|
20, 21 |
2385 |
|
NM_001324120.1 |
NP_001311049.1
|
20, 21 |
2564 |
|
NM_001324122.1 |
NP_001311051.1
|
21, 22 |
2585 |
|
NM_001324123.1 |
NP_001311052.1
|
20, 21 |
2464 |
|
NM_001324124.1 |
NP_001311053.1
|
19, 20 |
2347 |
|
NM_001324125.1 |
NP_001311054.1
|
17, 18 |
2146 |
|
NM_001324126.1 |
NP_001311055.1
|
19, 20 |
2335 |
|
NM_014608.4 |
NP_055423.1
|
20, 21 |
2437 |
|
XM_011543873.2 |
XP_011542175.1
|
20, 21 |
2786 |
|
XM_011543874.1 |
XP_011542176.1
|
20, 21 |
2819 |
|
XM_011543876.2 |
XP_011542178.2
|
20, 21 |
2387 |
|
XM_017022023.1 |
XP_016877512.1
|
20, 21 |
2792 |
|
XM_017022024.1 |
XP_016877513.1
|
20, 21 |
2800 |
|
GenBank mRNA |
AK125936.1 |
- |
19, 20 |
2490 |
|
AK127094.1 |
- |
5, 6 |
1285 |
|
AK291604.1 |
- |
20, 21 |
2364 |
|
AL357618.1 |
- |
4, 5 |
429 |
|
AY763577.1 |
- |
20, 21 |
2401 |
|
AY763578.1 |
- |
21, 22 |
2467 |
|
AY763579.1 |
- |
5, 6 |
1285 |
|
BC001306.2 |
- |
3, 4 |
253 |
|
BC005097.1 |
- |
20, 21 |
2328 |
3163591 |
D38549.1 |
- |
20, 21 |
2331 |
|
KJ534819.1 |
- |
19, 20 |
2276 |
|
View Full Product Details