Gene Transcripts

Gene Symbol
Klrb1
Entrez Gene ID
100043861
Gene Name
killer cell lectin-like receptor subfamily B member 1
Gene Aliases
4930431A04Rik, Gm4696, Klrb1g, Klrb6, Ly-55, Ly55, NKR-P1G, Nkrp-1e, Nkrp1g
Chromosome Location
Chr. 6: 128706093 - 128764561 on Build GRCm38
UniGene ID
Mm.66911
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099918.1 NP_001093388.1 3 204
GenBank mRNA DQ113419.1 - 3 204
HQ713436.1 - 3 204
JF429832.1 - 3 217

View Full Product Details