Gene Transcripts

Gene Symbol
Atr
Entrez Gene ID
245000
Gene Name
ataxia telangiectasia and Rad3 related
Gene Aliases
Chromosome Location
Chr. 9: 95857597 - 95952760 on Build GRCm38
UniGene ID
Mm.212462
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019864.1 NP_063917.1 15 3175
GenBank mRNA AK142244.1 - 15 3136
AK171072.1 - 15 3107

View Full Product Details