Gene Transcripts

Gene Symbol
Rhox2b
Entrez Gene ID
100039913
Gene Name
reproductive homeobox 2B
Gene Aliases
OTTMUSG00000017138, Rhox2.2, Rhox2a
Chromosome Location
Chr. X: 37411736 - 37416844 on Build GRCm38
UniGene ID
Mm.415046
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099316.1 NP_001092786.1 3 508
GenBank mRNA BC049546.1 - 508

Gene Symbol
Rhox2a
Entrez Gene ID
75199
Gene Name
reproductive homeobox 2A
Gene Aliases
4930539I12Rik, Rhox2, Rhox2.1
Chromosome Location
Chr. X: 37244402 - 37249690 on Build GRCm38
UniGene ID
Mm.391240
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029203.2 NP_083479.1 3 530
GenBank mRNA AK015997.1 - 3 531
BC132216.1 - 3 524 40130493
BC132220.1 - 3 524 40130497
BC145534.1 - 3 524
DQ058639.1 - 3 531

Gene Symbol
Rhox2c
Entrez Gene ID
100039948
Gene Name
reproductive homeobox 2C
Gene Aliases
OTTMUSG00000017149, Rhox2.3, Rhox2b
Chromosome Location
Chr. X: 37453480 - 37458417 on Build GRCm38
UniGene ID
Mm.477362
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099318.1 NP_001092788.1 530

Gene Symbol
Rhox2e
Entrez Gene ID
100040016
Gene Name
reproductive homeobox 2E
Gene Aliases
ENSMUSG00000063709, OTTMUSG00000017141, Rhox2.5, Rhox2d
Chromosome Location
Chr. X: 37530322 - 37541809 on Build GRCm38
UniGene ID
Mm.455498
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001085348.1 NP_001078817.1 530

View Full Product Details