Gene Transcripts
- Gene Symbol
- Schip1
- Entrez Gene ID
- 30953
- Gene Name
- schwannomin interacting protein 1
- Gene Aliases
- Nf2ip, Schip-1
- Chromosome Location
- Chr. 3: 68064802 - 68626482 on Build GRCm38
- UniGene ID
- Mm.331064
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001113420.1 |
NP_001106891.1
|
5, 6 |
765 |
|
NM_001113421.1 |
NP_001106892.1
|
6, 7 |
1449 |
|
NM_001282044.1 |
NP_001268973.1
|
6, 7 |
1432 |
|
NM_001282045.1 |
NP_001268974.1
|
6, 7 |
1290 |
|
NM_013928.5 |
NP_038956.2
|
5, 6 |
855 |
|
GenBank mRNA |
AF145716.1 |
- |
5, 6 |
961 |
|
AK021361.1 |
- |
5, 6 |
716 |
|
AK083464.1 |
- |
6, 7 |
1275 |
|
AK146930.1 |
- |
6, 7 |
1406 |
|
AK168005.1 |
- |
6, 7 |
1359 |
|
BC060529.1 |
- |
5, 6 |
789 |
|
EU163407.1 |
- |
6, 7 |
1449 |
|
EU163408.1 |
- |
5, 6 |
765 |
|
KM233715.1 |
- |
5, 6 |
711 |
|
KM233716.1 |
- |
5, 6 |
716 |
|
- Gene Symbol
- Gm21949
- Entrez Gene ID
- 100505386
- Gene Name
- predicted gene, 21949
- Gene Aliases
- Iqcj-schip1, Schip-1-iqcj, Schip1
- Chromosome Location
- Chr. 3: 67892220 - 68626482 on Build GRCm38
- UniGene ID
- Mm.331064
- Species
- Mus musculus
View Full Product Details