Gene Transcripts

Gene Symbol
Dtymk
Entrez Gene ID
21915
Gene Name
deoxythymidylate kinase
Gene Aliases
AU044245, CDC8, D10Ertd203e, TMPK, TYPK, Tmk, mtmk
Chromosome Location
Chr. 1: 93792576 - 93801934 on Build GRCm38
UniGene ID
Mm.250332
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105667.1 NP_001099137.1 2 249
NM_023136.2 NP_075625.1 2 249
GenBank mRNA AA014471.1 - 1 15
AK009220.1 - 2 222
AK011955.1 - 2 192
AK019284.1 - 2 184
AK019288.1 - 2 223
AK019445.1 - 2 219
AK088416.1 - 2 209
AK160747.1 - 2 219
AK210481.1 - 2 59
BC030178.1 - 2 211 5039765
BC069902.1 - 2 220 5026527
BY098372.1 - 2 250
CB182101.1 - 2 205
CF983443.1 - 2 237
S82852.1 - 2 190
S82853.1 - 2 190

View Full Product Details