Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001114087.2 | NP_001107559.1 | 7 | 652 | |
NM_001114088.2 | NP_001107560.1 | 7 | 652 | ||
NM_026131.4 | NP_080407.3 | 7 | 550 | ||
GenBank mRNA | AK003019.1 | - | 8 | 626 | |
AK003338.1 | - | 7 | 541 | ||
AK010141.1 | - | 7 | 539 | ||
AK010339.1 | - | 5 | 645 | ||
AK033091.1 | - | 644 | |||
AK049902.1 | - | 5 | 1350 | ||
AK077998.1 | - | 7 | 644 | ||
AK128987.1 | - | 7 | 640 | ||
AK142933.1 | - | 7 | 636 | ||
AK167476.1 | - | 7 | 644 | ||
AK184906.1 | - | 1 | 17 | ||
BC049565.1 | - | 7 | 538 | 6588160 | |
BC052698.1 | - | 7 | 623 |