Gene Transcripts
- Gene Symbol
- Rhox4b
- Entrez Gene ID
- 57737
- Gene Name
- reproductive homeobox 4B
- Gene Aliases
- 5430432L21Rik, Ehox, Rhox4.2, Rhox4a
- Chromosome Location
- Chr. X: 37432494 - 37437278 on Build GRCm38
- UniGene ID
- Mm.327419
- Species
- Mus musculus
- Gene Symbol
- Rhox4d
- Entrez Gene ID
- 664610
- Gene Name
- reproductive homeobox 4D
- Gene Aliases
- Rhox4.4, Rhox4c
- Chromosome Location
- Chr. X: 37514389 - 37519176 on Build GRCm38
- UniGene ID
- Mm.381921
- Species
- Mus musculus
- Gene Symbol
- Rhox4e
- Entrez Gene ID
- 194856
- Gene Name
- reproductive homeobox 4E
- Gene Aliases
- Rhox4, Rhox4.5, Rhox4d
- Chromosome Location
- Chr. X: 37557412 - 37562282 on Build GRCm38
- UniGene ID
- Mm.379498
- Species
- Mus musculus
- Gene Symbol
- Rhox4g
- Entrez Gene ID
- 664608
- Gene Name
- reproductive homeobox 4G
- Gene Aliases
- Rhox4.6, Rhox4.7, Rhox4f
- Chromosome Location
- Chr. X: 37646501 - 37651327 on Build GRCm38
- UniGene ID
- Mm.491809
- Species
- Mus musculus
View Full Product Details