Gene Transcripts
- Gene Symbol
- CALU
- Entrez Gene ID
- 813
- Gene Name
- calumenin
- Gene Aliases
- Chromosome Location
- Chr. 7: 128739292 - 128773423 on Build GRCh38
- UniGene ID
- Hs.743262
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001199671.1 |
NP_001186600.1
|
4, 5 |
1383 |
|
NM_001199673.1 |
NP_001186602.1
|
3, 4 |
568 |
|
NM_001219.4 |
NP_001210.1
|
3, 4 |
568 |
|
XM_017012659.1 |
XP_016868148.1
|
3, 4 |
933 |
|
GenBank mRNA |
AF013759.1 |
- |
3, 4 |
474 |
|
AK056338.1 |
- |
4, 5 |
1271 |
|
AK074862.1 |
- |
3, 4 |
518 |
|
AK312446.1 |
- |
3, 4 |
511 |
|
BC013383.2 |
- |
3, 4 |
467 |
3507961 |
BT006826.1 |
- |
2, 3 |
416 |
|
BT020143.1 |
- |
2, 3 |
416 |
|
CR457039.1 |
- |
2, 3 |
416 |
|
CR541835.1 |
- |
2, 3 |
416 |
|
HM002604.1 |
- |
4, 5 |
1384 |
|
HM002606.1 |
- |
3, 4 |
568 |
|
HM002607.1 |
- |
3, 4 |
568 |
|
HM002611.1 |
- |
3, 4 |
568 |
|
U67280.1 |
- |
3, 4 |
478 |
|
View Full Product Details