Gene Transcripts

Gene Symbol
Kifap3
Entrez Gene ID
289168
Gene Name
kinesin-associated protein 3
Gene Aliases
Chromosome Location
Chr. 13: 82072497 - 82217253 on Build Rnor_6.0
UniGene ID
Rn.163223
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105964.1 NP_001099434.1 15 2088
XM_008769623.1 XP_008767845.1 16 2220

View Full Product Details