Gene Transcripts

Gene Symbol
Wdr92
Entrez Gene ID
498418
Gene Name
WD repeat domain 92
Gene Aliases
RGD1563863
Chromosome Location
Chr. 14: 100373129 - 100394143 on Build Rnor_6.0
UniGene ID
Rn.25205
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127559.2 NP_001121031.1 3, 4 455
GenBank mRNA BC162042.1 - 3, 4 455

View Full Product Details