Gene Transcripts

Gene Symbol
Chtf8
Entrez Gene ID
364996
Gene Name
CTF8, chromosome transmission fidelity factor 8 homolog (S. cerevisiae)
Gene Aliases
Ctf8, RGD1306894
Chromosome Location
Chr. 19: 39077580 - 39087880 on Build Rnor_6.0
UniGene ID
Rn.3278
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001194951.1 NP_001181880.1 3 706
GenBank mRNA BC166818.1 - 1 359

View Full Product Details