Gene Transcripts

Gene Symbol
Cryge
Entrez Gene ID
24279
Gene Name
crystallin, gamma E
Gene Aliases
Cryg5, Len
Chromosome Location
Chr. 9: 71764317 - 71767411 on Build Rnor_6.0
UniGene ID
Rn.44578
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173289.1 NP_775411.1 3 322
XM_008767178.1 XP_008765400.1 4 499
GenBank mRNA J00716.1 - 280

Gene Symbol
Crygf
Entrez Gene ID
689947
Gene Name
crystallin, gamma F
Gene Aliases
Cryg6, Len
Chromosome Location
Chr. 9: 72933304 - 72935468 on Build Rnor_6.0
UniGene ID
Rn.102739
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109557.1 NP_001103027.1 3 375

View Full Product Details