Gene Transcripts

Gene Symbol
Trex1
Entrez Gene ID
100049583
Gene Name
three prime repair exonuclease 1
Gene Aliases
RGD1309596
Chromosome Location
Chr. 8: 117796127 - 117797427 on Build Rnor_6.0
UniGene ID
Rn.98617
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024989.1 NP_001020160.1 2 547
GenBank mRNA BC091242.1 - 2 547
FQ233170.1 - 2 597

View Full Product Details