Gene Transcripts

Gene Symbol
Dexi
Entrez Gene ID
497857
Gene Name
dexamethasone-induced transcript
Gene Aliases
RGD1564938
Chromosome Location
Chr. 10: 5199226 - 5211913 on Build Rnor_6.0
UniGene ID
Rn.15447
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109026.1 NP_001102496.1 1 534

View Full Product Details