Gene Transcripts

Gene Symbol
Lman2l
Entrez Gene ID
301343
Gene Name
lectin, mannose-binding 2-like
Gene Aliases
Chromosome Location
Chr. 9: 43020940 - 43023015 on Build Rnor_6.0
UniGene ID
Rn.102571
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106900.1 NP_001100370.1 2 243
GenBank mRNA BC161950.1 - 227

View Full Product Details