Gene Transcripts

Gene Symbol
Ost4
Entrez Gene ID
100188932
Gene Name
oligosaccharyltransferase 4 homolog (S. cerevisiae)
Gene Aliases
Chromosome Location
Chr. 6: 26836216 - 26837440 on Build Rnor_6.0
UniGene ID
Rn.34416
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134690.1 NP_001128162.1 2 405
NM_001134691.1 NP_001128163.1 3 305
GenBank mRNA BC157814.1 - 2 383
CB706964.1 - 3 266

View Full Product Details