Gene Transcripts

Gene Symbol
Pdxp
Entrez Gene ID
727679
Gene Name
pyridoxal (pyridoxine, vitamin B6) phosphatase
Gene Aliases
Rbp1
Chromosome Location
Chr. 7: 120140460 - 120145908 on Build Rnor_6.0
UniGene ID
Rn.222285
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135819.1 NP_001129291.1 2 1788
GenBank mRNA AF318578.1 - 2 1441
BC166563.1 - 2 1788 7316162

View Full Product Details