Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001278731.1 | NP_001265660.1 | 10 | 1430 | |
NM_001278732.1 | NP_001265661.1 | 9 | 1383 | ||
NM_012208.3 | NP_036340.1 | 11 | 1505 | ||
XM_011537619.1 | XP_011535921.1 | 12 | 1518 | ||
XM_011537620.1 | XP_011535922.1 | 11 | 1419 | ||
XM_017009287.1 | XP_016864776.1 | 10 | 1406 | ||
XM_017009288.1 | XP_016864777.1 | 12 | 1569 | ||
XM_017009289.1 | XP_016864778.1 | 12 | 1562 | ||
XM_017009290.1 | XP_016864779.1 | 11 | 1629 | ||
XM_017009291.1 | XP_016864780.1 | 11 | 1322 | ||
XM_017009292.1 | XP_016864781.1 | 11 | 1266 | ||
GenBank mRNA | AK293269.1 | - | 9 | 1163 | |
AK293390.1 | - | 10 | 1247 | ||
AK298661.1 | - | 6 | 801 | ||
AK299971.1 | - | 8 | 959 | ||
AK312976.1 | - | 11 | 1323 | ||
BC007680.2 | - | 11 | 1324 | 3628006 | |
BC014982.1 | - | 11 | 1469 | 4859849 | |
U18937.1 | - | 11 | 1431 |