Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001278731.1 | NP_001265660.1 | 4 | 644 | |
NM_012208.3 | NP_036340.1 | 5 | 719 | ||
XM_011537619.1 | XP_011535921.1 | 6 | 732 | ||
XM_011537620.1 | XP_011535922.1 | 6 | 732 | ||
XM_017009287.1 | XP_016864776.1 | 5 | 719 | ||
XM_017009288.1 | XP_016864777.1 | 6 | 783 | ||
XM_017009289.1 | XP_016864778.1 | 6 | 776 | ||
XM_017009290.1 | XP_016864779.1 | 5 | 769 | ||
XM_017009291.1 | XP_016864780.1 | 5 | 462 | ||
XM_017009292.1 | XP_016864781.1 | 5 | 406 | ||
GenBank mRNA | AK293390.1 | - | 4 | 461 | |
AK299971.1 | - | 3 | 272 | ||
AK309832.1 | - | 5 | 817 | ||
AK311277.1 | - | 5 | 815 | ||
AK312976.1 | - | 5 | 537 | ||
BC007680.2 | - | 5 | 538 | 3628006 | |
BC014982.1 | - | 5 | 683 | 4859849 | |
U18937.1 | - | 5 | 645 |