Gene Transcripts

Gene Symbol
TSPAN15
Entrez Gene ID
23555
Gene Name
tetraspanin 15
Gene Aliases
2700063A19Rik, NET-7, NET7, TM4SF15
Chromosome Location
Chr. 10: 69451464 - 69549508 on Build GRCh38
UniGene ID
Hs.499941
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012339.3 NP_036471.1 4 508
XM_005269666.4 XP_005269723.1 2 968
XM_005269667.3 XP_005269724.1 2 251
XM_011539562.2 XP_011537864.1 2 211
XM_011539563.2 XP_011537865.1 2 422
XM_017016010.1 XP_016871499.1 4 514
GenBank mRNA AF120266.1 - 4 499
BC003157.1 - 4 502 2958221
BC004161.2 - 4 502 2958221

View Full Product Details