Gene Transcripts

Gene Symbol
CCNDBP1
Entrez Gene ID
23582
Gene Name
cyclin D1 binding protein 1
Gene Aliases
DIP1, GCIP, HHM
Chromosome Location
Chr. 15: 43185118 - 43197177 on Build GRCh38
UniGene ID
Hs.36794
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012142.4 NP_036274.3 8 847
NR_027513.2 6 670
NR_027514.2 7 750
NR_045998.1 7 767
NR_045999.1 5 610
XM_006720448.1 XP_006720511.1 6 769
GenBank mRNA AF082569.1 - 8 706
AF087852.1 - 8 692
AF113535.1 - 6 656
AF132034.1 - 8 662
AF246144.1 - 8 664
AK075146.1 - 8 676
AK075296.1 - 6 661
AK124193.1 - 3 1251
AK128849.1 - 4 1891
AK225143.1 - 8 662
AK225645.1 - 8 676
AK290665.1 - 8 676
AK290707.1 - 7 629
AK295017.1 - 7 579
AK308817.1 - 7 1271
AL552334.3 - 8 786
BC009689.1 - 8 690 3896421
BQ439439.1 - 5 424
CR450331.1 - 8 616

View Full Product Details