Gene Transcripts

Gene Symbol
EPHB4
Entrez Gene ID
2050
Gene Name
EPH receptor B4
Gene Aliases
HTK, MYK1, TYRO11
Chromosome Location
Chr. 7: 100802565 - 100827521 on Build GRCh38
UniGene ID
Hs.437008
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004444.4 NP_004435.3 17 3956
XM_017011816.1 XP_016867305.1 17 3613
GenBank mRNA AB209644.1 - 16 4666
AY056048.1 - 16 3394
BC004264.1 - 14 3041
BC052804.1 - 17 3633 6584384
U07695.1 - 17 3550

View Full Product Details