Gene Transcripts

Gene Symbol
RAB26
Entrez Gene ID
25837
Gene Name
RAB26, member RAS oncogene family
Gene Aliases
V46133
Chromosome Location
Chr. 16: 2148144 - 2154165 on Build GRCh38
UniGene ID
Hs.3797
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001308053.1 NP_001294982.1 5 556
NM_014353.4 NP_055168.2 4 517
XM_011522448.1 XP_011520750.1 4 543
XM_011522450.2 XP_011520752.1 3 1334
GenBank mRNA AB027137.1 - 3 188
AF498952.1 - 3 185
AK308560.1 - 6 613
AK309666.1 - 5 521
AK314110.1 - 5 556
AY646153.1 - 4 534
BC066913.1 - 4 523 5262795

View Full Product Details