Gene Transcripts

Gene Symbol
MOB4
Entrez Gene ID
25843
Gene Name
MOB family member 4, phocein
Gene Aliases
2C4D, CGI-95, MOB1, MOB3, MOBKL3, PHOCN, PREI3
Chromosome Location
Chr. 2: 197515571 - 197553699 on Build GRCh38
UniGene ID
Hs.633165
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100819.2 NP_001094289.1 6 469
NM_001204094.1 NP_001191023.1 7 486
NM_015387.4 NP_056202.2 7 532
NM_199482.3 NP_955776.1 7 542
GenBank mRNA AB015441.1 - 7 491
AF093825.1 - 7 507
AF151853.1 - 7 507
AF250319.1 - 7 517
AJ580638.1 - 7 488
AK027043.1 - 7 529
AK289377.1 - 7 522
AK292938.1 - 7 528
AK297294.1 - 403
AK297514.1 - 6 428
AK307497.1 - 6 400
AL080070.1 - 7 501
BC005237.1 - 7 532 3996585
CR457371.1 - 7 488
DC298610.1 - 7 542
DC350915.1 - 6 465

Gene Symbol
HSPE1-MOB4
Entrez Gene ID
100529241
Gene Name
HSPE1-MOB4 readthrough
Gene Aliases
HSPE1-PHOCN
Chromosome Location
Chr. 2: 197499997 - 197553699 on Build GRCh38
UniGene ID
Hs.633165
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001202485.1 NP_001189414.1 8 1036
GenBank mRNA CR990871.1 - 8 620

View Full Product Details