Gene Transcripts
- Gene Symbol
- SUMF2
- Entrez Gene ID
- 25870
- Gene Name
- sulfatase modifying factor 2
- Gene Aliases
- pFGE
- Chromosome Location
- Chr. 7: 56064007 - 56080672 on Build GRCh38
- UniGene ID
- Hs.279696
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001042469.1 |
NP_001035934.2
|
7, 8 |
872 |
|
NM_001042470.1 |
NP_001035935.2
|
5, 6 |
665 |
|
NM_001146333.1 |
NP_001139805.1
|
7, 8 |
760 |
|
NM_015411.2 |
NP_056226.2
|
8, 9 |
917 |
|
XM_017011939.1 |
XP_016867428.1
|
9, 10 |
1035 |
|
XM_017011940.1 |
XP_016867429.1
|
8, 9 |
1207 |
|
GenBank mRNA |
AK075477.1 |
- |
8, 9 |
876 |
|
AK297042.1 |
- |
7, 8 |
788 |
|
AK297793.1 |
- |
9 |
909 |
|
AK298605.1 |
- |
6, 7 |
650 |
|
AK300488.1 |
- |
7, 8 |
719 |
|
AK301627.1 |
- |
5, 6 |
582 |
|
AL050037.1 |
- |
6, 7 |
637 |
|
AY323911.1 |
- |
8, 9 |
829 |
|
AY359103.1 |
- |
8, 9 |
847 |
|
BC000224.1 |
- |
7, 8 |
652 |
|
BC006159.1 |
- |
8, 9 |
815 |
|
BC065222.1 |
- |
5, 6 |
553 |
|
BC084539.1 |
- |
8, 9 |
847 |
6599080 |
BC111092.1 |
- |
8, 9 |
870 |
40068651 |
BP256067.1 |
- |
6, 7 |
559 |
|
CR936749.1 |
- |
7, 8 |
807 |
|
CR936757.1 |
- |
8, 9 |
860 |
|
KU178583.1 |
- |
7, 8 |
742 |
|
KU178584.1 |
- |
6, 7 |
697 |
|
KU178585.1 |
- |
4, 5 |
490 |
|
View Full Product Details