Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015500.1 | NP_056315.1 | 5 | 946 | |
NM_199050.2 | NP_950251.1 | 4 | 445 | ||
XM_005261109.4 | XP_005261166.1 | 4 | 486 | ||
XM_011529522.1 | XP_011527824.1 | 5 | 1046 | ||
XM_011529523.2 | XP_011527825.1 | 6 | 1712 | ||
XM_011529524.2 | XP_011527826.1 | 5 | 1046 | ||
XM_017028315.1 | XP_016883804.1 | 2 | 182 | ||
GenBank mRNA | AB047784.1 | - | 5 | 946 | |
AB047785.1 | - | 5 | 946 | ||
AK128550.1 | - | 2 | 285 | ||
AL833323.1 | - | 4 | 445 | ||
BC062323.1 | - | 4 | 427 | 5419622 |