Gene Transcripts

Gene Symbol
RNF11
Entrez Gene ID
26994
Gene Name
ring finger protein 11
Gene Aliases
CGI-123, SID1669
Chromosome Location
Chr. 1: 51236273 - 51273447 on Build GRCh38
UniGene ID
Hs.309641
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014372.4 NP_055187.1 2 738
GenBank mRNA AB024703.1 - 2 381
AF151881.1 - 2 381
AK293047.1 - 4 567
AK313140.1 - 2 500
BC020964.2 - 2 333 3917224
BC047654.1 - 2 706 5261966

View Full Product Details