Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015697.7 | NP_056512.5 | 4 | 717 | |
XM_011531855.2 | XP_011530157.1 | 4 | 928 | ||
XM_011531857.2 | XP_011530159.1 | 4 | 926 | ||
XM_011531859.2 | XP_011530161.1 | 4 | 925 | ||
XM_011531860.2 | XP_011530162.1 | 4 | 925 | ||
XM_011531862.2 | XP_011530164.1 | 4 | 924 | ||
XM_011531863.2 | XP_011530165.1 | 4 | 921 | ||
XM_011531866.2 | XP_011530168.1 | 4 | 917 | ||
XM_011531867.2 | XP_011530169.1 | 4 | 840 | ||
XM_017008031.1 | XP_016863520.1 | 4 | 840 | ||
XM_017008032.1 | XP_016863521.1 | 2 | 110 | ||
GenBank mRNA | AF091086.1 | - | 3 | 266 | |
AJ621061.3 | - | 4 | 717 | ||
AK129583.1 | - | 4 | 473 | ||
AK300972.1 | - | 4 | 598 | ||
BC008804.2 | - | 4 | 574 | 3954787 | |
BC020728.1 | - | 4 | 614 | 4696019 | |
BG699931.1 | - | 4 | 603 |