Gene Transcripts

Gene Symbol
SNX24
Entrez Gene ID
28966
Gene Name
sorting nexin 24
Gene Aliases
PRO1284, SBBI31
Chromosome Location
Chr. 5: 122845465 - 123029318 on Build GRCh38
UniGene ID
Hs.483200
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014035.2 NP_054754.1 4, 5 507
XM_005271972.4 XP_005272029.1 4, 5 396
XM_006714592.3 XP_006714655.1 4, 5 462
XM_011543349.2 XP_011541651.1 5, 6 527
XM_011543350.2 XP_011541652.1 5, 6 526
XM_017009394.1 XP_016864883.1 4, 5 332
GenBank mRNA AF139461.1 - 4, 5 396
AY044655.1 - 4, 5 338
AY358098.1 - 4, 5 356
BC010886.1 - 4, 5 467 3538426
BC069012.1 - 4, 5 454 4585781
BQ278046.1 - 4, 5 479

View Full Product Details